Anti-NMS Rabbit Polyclonal Antibody (Rhodamine)
Lieferant: US Biological
N2171-79U7
USBIN2171-79U7EA 1160 EUR
USBIN2171-79U7
Anti-NMS Rabbit Polyclonal Antibody (Rhodamine)
Antikörper
Type: Primary
Antigen: NMS (neuromedin S)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse