Anti-Neuropeptide Y6 Receptor Rabbit Polyclonal Antibody
Lieferant: US Biological
N2176-67
USBIN2176-67EA 742 EUR
USBIN2176-67
Anti-Neuropeptide Y6 Receptor Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.5-1 ?g/ml
Type: Primary
Antigen: Neuropeptide Y6 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse