Anti-Niban Like Protein 1 Rabbit Polyclonal Antibody

Lieferant: US Biological
N2497-90
USBIN2497-90EA 380 EUR
USBIN2497-90
Anti-Niban Like Protein 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:1000; Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: Niban Like Protein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR