Anti-Nicotinic Acetylcholine Receptor alpha 1 Rabbit Polyclonal Antibody

Lieferant: US Biological
N2562-08D
USBIN2562-08DEA 639 EUR
USBIN2562-08D
Anti-Nicotinic Acetylcholine Receptor alpha 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.75 ?g/ml

Type: Primary
Antigen: Nicotinic Acetylcholine Receptor alpha 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR