Anti-Nicotinic Acetylcholine Receptor alpha 4 Rabbit Polyclonal Antibody

Lieferant: US Biological
N2563-01
USBIN2563-01EA 754 EUR
USBIN2563-01
Anti-Nicotinic Acetylcholine Receptor alpha 4 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Nicotinic Acetylcholine Receptor alpha 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR