Anti-Nicotinic Receptor, alpha 4 Rabbit Polyclonal Antibody
Lieferant: US Biological
N2565-21A
USBIN2565-21AEA 634 EUR
USBIN2565-21A
Anti-Nicotinic Receptor, alpha 4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nicotinic Receptor, alpha 4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen