Anti-Nicotinic Receptor, alpha 5 Rabbit Polyclonal Antibody

Lieferant: US Biological
N2565-25E
USBIN2565-25EEA 512 EUR
USBIN2565-25E
Anti-Nicotinic Receptor, alpha 5 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:8

Type: Primary
Antigen: Nicotinic Receptor, alpha 5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR