Anti-Nicotinic Receptor, alpha 7 Rabbit Polyclonal Antibody
Lieferant: US Biological
N2565-26D
USBIN2565-26DEA 633 EUR
USBIN2565-26D
Anti-Nicotinic Receptor, alpha 7 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 25 ?g/ml; Immunochemistry: 20 ?g/ml
Type: Primary
Antigen: Nicotinic Receptor, alpha 7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen