Anti-Nicotinic Receptor, alpha 7 Rabbit Polyclonal Antibody
Lieferant: US Biological
N2565-26E
USBIN2565-26EEA 613 EUR
USBIN2565-26E
Anti-Nicotinic Receptor, alpha 7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nicotinic Receptor, alpha 7
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen