Anti-Nicotinic Receptor, beta 2 Rabbit Polyclonal Antibody
Lieferant: US Biological
N2565-30A
USBIN2565-30AEA 410 EUR
USBIN2565-30A
Anti-Nicotinic Receptor, beta 2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:200; Immunochemistry: 1:400
Type: Primary
Antigen: Nicotinic Receptor, beta 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen