Anti-Nicotinic Receptor, beta 2 Rabbit Polyclonal Antibody
Lieferant: US Biological
N2565-30E
USBIN2565-30EEA 613 EUR
USBIN2565-30E
Anti-Nicotinic Receptor, beta 2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nicotinic Receptor, beta 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen