Anti-Nicotinic Receptor, beta 3 Rabbit Polyclonal Antibody

Lieferant: US Biological
N2565-32A
USBIN2565-32AEA 634 EUR
USBIN2565-32A
Anti-Nicotinic Receptor, beta 3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Nicotinic Receptor, beta 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR