Anti-Nicotinic Receptor, beta 3 Rabbit Polyclonal Antibody

Lieferant: US Biological
N2565-32B
USBIN2565-32BEA 512 EUR
USBIN2565-32B
Anti-Nicotinic Receptor, beta 3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:100; Immunochemistry: 1:50

Type: Primary
Antigen: Nicotinic Receptor, beta 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR