Anti-Nitrofurantoin Sheep Polyclonal Antibody

Lieferant: US Biological
N2593
USBIN2593EA 1020 EUR
USBIN2593
Anti-Nitrofurantoin Sheep Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 5.0 ?g/ml

Type: Primary
Antigen: Nitrofurantoin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR