Anti-NMDA 1 Receptor, C2’ Splice Variant Rabbit Polyclonal Antibody

Lieferant: US Biological
N2904-11
USBIN2904-11EA 857 EUR
USBIN2904-11
Anti-NMDA 1 Receptor, C2’ Splice Variant Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NMDA 1 Receptor, C2’ Splice Variant
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR