Anti-NOS1 Rabbit Polyclonal Antibody

Lieferant: US Biological
N2950-01D
USBIN2950-01DEA 513 EUR
USBIN2950-01D
Anti-NOS1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:2000; Immunochemistry: 1:1000-1:5000

Type: Primary
Antigen: NOS1 (nitric oxide synthase 1 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR