Anti-Noradrenaline Rabbit Polyclonal Antibody (Unconjugated)
Lieferant: US Biological
N3950-04
USBIN3950-04EA 795 EUR
USBIN3950-04
Anti-Noradrenaline Rabbit Polyclonal Antibody (Unconjugated)
Antikörper
Type: Primary
Antigen: Noradrenaline
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen