Anti-19-Nortestosterone Sheep Polyclonal Antibody

Lieferant: US Biological
N4004
USBIN4004EA 695 EUR
USBIN4004
Anti-19-Nortestosterone Sheep Polyclonal Antibody
Antikörper
Recommended dilution: ELISA:10 ?g/ml

Type: Primary
Antigen: 19-Nortestosterone
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR