Anti-Novel Organic Cation Transporter 2 Rabbit Polyclonal Antibody

Lieferant: US Biological
N5376-54
USBIN5376-54EA 708 EUR
USBIN5376-54
Anti-Novel Organic Cation Transporter 2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:5000; ELISA: 1:10000-1:50000

Type: Primary
Antigen: Novel Organic Cation Transporter 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR