Anti-Novel Organic Cation Transporter 2 Rabbit Polyclonal Antibody

Lieferant: US Biological
N5376-54A-100
USBIN5376-54A-100EA 742 EUR
USBIN5376-54A-100
Anti-Novel Organic Cation Transporter 2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-10 ?g/ml; ELISA: 0.5-1.0 ?g/ml

Type: Primary
Antigen: Novel Organic Cation Transporter 2
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR