Anti-NR0B11 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: US Biological
N6889-92B7FITC
USBIN6889-92B7FITCEA 884 EUR
USBIN6889-92B7FITC
Anti-NR0B11 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Primary
Antigen: NR0B11
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR