Anti-Nuclear Receptor Subfamily 2 Group E Member 1 Mouse Monoclonal Antibody [clone: 9G408]
Lieferant: US Biological
N6889-94E2
USBIN6889-94E2EA 601 EUR
USBIN6889-94E2
Anti-Nuclear Receptor Subfamily 2 Group E Member 1 Mouse Monoclonal Antibody [clone: 9G408]
Antikörper
Type: Primary
Antigen: Nuclear Receptor Subfamily 2 Group E Member 1
Clonality: monoclonal
Clone: 9G408
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat