Anti-NME1 Mouse Monoclonal Antibody [clone: 10K87]

Lieferant: US Biological
N7100-61
USBIN7100-61EA 802 EUR
USBIN7100-61
Anti-NME1 Mouse Monoclonal Antibody [clone: 10K87]
Antikörper
Recommended dilution: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: NME1 (NME/NM23 nucleoside diphosphate kinase 1)
Clonality: monoclonal
Clone: 10K87
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR