Anti-Nucleoside Diphosphate Kinase D Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
N7100-63N-BIOT
USBIN7100-63N-BIOTEA 884 EUR
USBIN7100-63N-BIOT
Anti-Nucleoside Diphosphate Kinase D Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Nucleoside Diphosphate Kinase D
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR