Anti-Nucleoside Diphosphate Kinase A Chicken Polyclonal Antibody

Lieferant: US Biological
N7100-65
USBIN7100-65EA 601 EUR
USBIN7100-65
Anti-Nucleoside Diphosphate Kinase A Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:200;

Type: Primary
Antigen: Nucleoside Diphosphate Kinase A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR