Anti-SLCO1B3, Solute Carrier Organic Anion Transporter Family, Member 1B3 Goat Polyclonal Antibody

Lieferant: US Biological
O3001-20
USBIO3001-20EA 640 EUR
USBIO3001-20
Anti-SLCO1B3, Solute Carrier Organic Anion Transporter Family, Member 1B3 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1: 2000

Type: Primary
Antigen: SLCO1B3, Solute Carrier Organic Anion Transporter Family, Member 1B3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR