Anti-Omentin Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
O6250-02B
USBIO6250-02BEA 1160 EUR
USBIO6250-02B
Anti-Omentin Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Omentin
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR