Anti-OSR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: US Biological
O8054-09-PE
USBIO8054-09-PEEA 884 EUR
USBIO8054-09-PE
Anti-OSR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000 Dot Blot: 1:500
Do not freeze!
Type: Primary
Antigen: OSR1 (odd-skipped related transciption factor 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human