Anti-OSTN Rabbit Polyclonal Antibody (Rhodamine)

Lieferant: US Biological
O8061-18G5
USBIO8061-18G5EA 1160 EUR
USBIO8061-18G5
Anti-OSTN Rabbit Polyclonal Antibody (Rhodamine)
Antikörper
Type: Primary
Antigen: OSTN (osteocrin)
Clonality: polyclonal
Clone:
Conjugation: Rhodamine
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR