Anti-p21,WAF1 Mouse Monoclonal Antibody [clone: 5F185]

Lieferant: US Biological
P1000-54A
USBIP1000-54AEA 754 EUR
USBIP1000-54A
Anti-p21,WAF1 Mouse Monoclonal Antibody [clone: 5F185]
Antikörper
Recommended dilution: Immunochemistry: 1:5

Type: Primary
Antigen: p21,WAF1
Clonality: monoclonal
Clone: 5F185
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mammalian, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR