Anti-p21,WAF1 Mouse Monoclonal Antibody [clone: 0.N.489]

Lieferant: US Biological
P1000-55-250
USBIP1000-55-250EA 361 EUR
USBIP1000-55-250
Anti-p21,WAF1 Mouse Monoclonal Antibody [clone: 0.N.489]
Antikörper
Type: Primary
Antigen: p21,WAF1
Clonality: monoclonal
Clone: 0.N.489
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR