Anti-p21Cip1/WAF1 Rabbit Polyclonal Antibody
Lieferant: US Biological
P1000-59Z
USBIP1000-59ZEA 380 EUR
USBIP1000-59Z
Anti-p21Cip1/WAF1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry:1:50-1:100
Type: Primary
Antigen: p21Cip1/WAF1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human