Anti-p21Cip1/WAF1 Rabbit Polyclonal Antibody

Lieferant: US Biological
P1000-59Z1
USBIP1000-59Z1EA 380 EUR
USBIP1000-59Z1
Anti-p21Cip1/WAF1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot:1:500-1:1000

Type: Primary
Antigen: p21Cip1/WAF1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR