Anti-PEG10 Mouse Monoclonal Antibody [clone: 1B1C4]

Lieferant: US Biological
P1800-80
USBIP1800-80EA 633 EUR
USBIP1800-80
Anti-PEG10 Mouse Monoclonal Antibody [clone: 1B1C4]
Antikörper
Recommended dilution: Immunofluorescence: 1:200-1:1000; ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200

Type: Primary
Antigen: PEG10 (paternally expressed 10)
Clonality: monoclonal
Clone: 1B1C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR