Anti-PEG10 Mouse Monoclonal Antibody [clone: 4C10A7]

Lieferant: US Biological
P1800-80A
USBIP1800-80AEA 633 EUR
USBIP1800-80A
Anti-PEG10 Mouse Monoclonal Antibody [clone: 4C10A7]
Antikörper
Recommended dilution: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunochemistry: 1:200-1:1000

Type: Primary
Antigen: PEG10 (paternally expressed 10)
Clonality: monoclonal
Clone: 4C10A7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR