Anti-PPY Rabbit Polyclonal Antibody

Lieferant: US Biological
P3000-04B
USBIP3000-04BEA 730 EUR
USBIP3000-04B
Anti-PPY Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:1000-1:2000

Type: Primary
Antigen: PPY (pancreatic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR