Anti-PPY Rabbit Polyclonal Antibody

Lieferant: US Biological
P3000-10H
USBIP3000-10HEA 390 EUR
USBIP3000-10H
Anti-PPY Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPY (pancreatic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR