Anti-PPY Goat Polyclonal Antibody
Lieferant: US Biological
P3000-10K
USBIP3000-10KEA 897 EUR
USBIP3000-10K
Anti-PPY Goat Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 5-15 ?g/ml
Type: Primary
Antigen: PPY (pancreatic polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human