Anti-Pantothenate Kinase 2, Mitochondrial Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
P3048-71N-BIOT
USBIP3048-71N-BIOTEA 884 EUR
USBIP3048-71N-BIOT
Anti-Pantothenate Kinase 2, Mitochondrial Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Pantothenate Kinase 2, Mitochondrial
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR