Anti-Paracetamol Sheep Polyclonal Antibody
Lieferant: US Biological
P3106-05B
USBIP3106-05BEA 614 EUR
USBIP3106-05B
Anti-Paracetamol Sheep Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Paracetamol
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen