Anti-PARP1 Rabbit monoclonal antibody Agarose [clone: 10g87]
Lieferant: US Biological
P3113-01Q
USBIP3113-01QEA 614 EUR
USBIP3113-01Q
Anti-PARP1 Rabbit monoclonal antibody Agarose [clone: 10g87]
Antikörper
Type: Primary
Antigen: PARP1 (poly (ADP-ribose) polymerase 1)
Clonality: monoclonal
Clone: 10g87
Conjugation: Agarose
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse, Rat