Anti-Peanut Protein Chicken Polyclonal Antibody
Lieferant: US Biological
P3129-87
USBIP3129-87EA 485 EUR
USBIP3129-87
Anti-Peanut Protein Chicken Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 2-5 ?g/ml
Type: Primary
Antigen: Peanut Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen