Anti-Penicillin Mouse Monoclonal Antibody [clone: Pen 9]

Lieferant: US Biological
P3175-02B
USBIP3175-02BEA 843 EUR
USBIP3175-02B
Anti-Penicillin Mouse Monoclonal Antibody [clone: Pen 9]
Antikörper
Type: Primary
Antigen: Penicillin
Clonality: monoclonal
Clone: Pen 9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR