Anti-Pepsin Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
P3270-07B
USBIP3270-07BEA 608 EUR
USBIP3270-07B
Anti-Pepsin Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Pepsin
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR