Anti-PRDX1 Rabbit Polyclonal Antibody

Lieferant: US Biological
P3351-95A
USBIP3351-95AEA 667 EUR
USBIP3351-95A
Anti-PRDX1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: PRDX1 (peroxiredoxin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR