Anti-PI3KC3 Rabbit Polyclonal Antibody (Biotin)
Lieferant: US Biological
P4073-33N-BIOT
USBIP4073-33N-BIOTEA 884 EUR
USBIP4073-33N-BIOT
Anti-PI3KC3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: PI3KC3
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human