Anti-Phospholipase D Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
P4074-51B
USBIP4074-51BEA 608 EUR
USBIP4074-51B
Anti-Phospholipase D Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Phospholipase D
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR