Anti-Phosphorylase B Sheep Polyclonal Antibody (Biotin)

Lieferant: US Biological
P4075-50D
USBIP4075-50DEA 608 EUR
USBIP4075-50D
Anti-Phosphorylase B Sheep Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Phosphorylase B
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR