Anti-Phosphorylase A Sheep Polyclonal Antibody (Biotin)

Lieferant: US Biological
P4075-88B
USBIP4075-88BEA 608 EUR
USBIP4075-88B
Anti-Phosphorylase A Sheep Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Phosphorylase A
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR