Anti-PI3KC2G Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
P4186-23F-BIOT
USBIP4186-23F-BIOTEA 884 EUR
USBIP4186-23F-BIOT
Anti-PI3KC2G Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: PI3KC2G
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR